Orders LinkContactUsLinkComponent
Accessories for microcentrifuges, mySPIN™
Accessories for microcentrifuges, mySPIN™
16×0,2 ml fixed angle tube rotor (singles or 2×8 strip) reinforced glass fiber, max. RCF 2000×g, max. speed 6000 min⁻¹ for microcentrifuge mySPIN™ 6
Catalog # HERA75004063
Supplier:   Thermo Scientific
Accessories for microcentrifuges, mySPIN™
Catalog # HERA75004063
Supplier:  Thermo Scientific
Accessories for microcentrifuges, mySPIN™
Accessories for microcentrifuges, mySPIN™
16×0,2 ml fixed angle tube rotor (singles or 2×8 strip) reinforced glass fiber, max. RCF 2000×g, max. speed 6000 min⁻¹ for microcentrifuge mySPIN™ 6
Price
63,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    16×0,2 ml fixed angle tube rotor (singles or 2×8 strip) reinforced glass fiber, max. RCF 2000×g, max. speed 6000 min⁻¹ for microcentrifuge mySPIN™ 6
  • Seq:
    02