Order Entry
Export
Orders LinkContactUsLinkComponent
Temperature sensors

Temperature sensors

Temperature probe, PT100 with two wire and cable (1,8 m), +250…+500 °C, 4×50 mm
Catalog # WINKWFMP1250-XX050
Supplier:   WINKLER
Temperature sensors
Catalog # WINKWFMP1250-XX050
Supplier:  WINKLER
Temperature sensors
Temperature sensors

Temperature probe, PT100 with two wire and cable (1,8 m), +250…+500 °C, 4×50 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Range:
    +250…+500 °C
  • Description:
    Temperature probe, PT100 with two wire and cable (1,8 m)
  • Ø×L:
    4×50 mm