Orders LinkContactUsLinkComponent
Inoculating loops, PS, calibrated
Inoculating loop, PS, sterile, 10 µl, Blue
  DELT302753
 :   DELTALAB
Inoculating loops, PS, calibrated
  DELT302753
 :  DELTALAB
Inoculating loops, PS, calibrated
Inoculating loop, PS, sterile, 10 µl, Blue
258,00 €
1 * 1.200 items
These calibrated, dual use inoculating loops offer a loop at one end to facilitate microbiological streaking, and a fine tip at the other specially designed for counting colonies.
  • High robustness and flexible
  • Colour-coded for ease of size identification
  • Eliminates the need for flaming
  • Sterilised by radiation
  • Capacity:
    10 µl
  • Colour:
    Blue
  • Description:
    Inoculating loop, PS, sterile