Orders LinkContactUsLinkComponent
Storage cabinets, full length
Storage cabinets, full length
Full length cabinet on leveller legs, with 6 shelves, 2, 1200×600×2100 mm
Catalog # 132-0993
Supplier:   Palbam
Storage cabinets, full length
Catalog # 132-0993
Supplier:  Palbam
Storage cabinets, full length
Storage cabinets, full length
Full length cabinet on leveller legs, with 6 shelves, 2, 1200×600×2100 mm
Price
9 350,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 304 with electropolished finish.
  • Removable shelves and sloping roof design as standard
  • Available on either castors or leveller legs
  • Assembly is quick and easy, requiring no special tools or expertise
Packaging: Typically available from stock for direct shipment from the supplier. Designed to be shipped flat-packed for self-assembly. Assembly instructions are included with all cabinets.
Specifications
  • Description:
    Full length cabinet on leveller legs, with 6 shelves
  • Seq:
    03
  • W×D×H:
    1200×600×2100 mm
  • Doors:
    2