Nessler reagent, tablets
DPD 3, 250
Catalog # LOVI511251BT
Supplier: Lovibond
Jump to:
Product Details & Documents
Synonyms: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
- CAS Number:7783-33-7
- MDL Number:MFCD00049656
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:250
- Pk Info:DPD 3



