Order Entry
Export
Orders LinkContactUsLinkComponent
Oleic acid

Oleic acid

25 L
Catalog # APPC142659.0716
Irritant
Supplier:   PanReac AppliChem
Oleic acid
Catalog # APPC142659.0716
Supplier:  PanReac AppliChem
CAS Number:  112-80-1
Oleic acid
Oleic acid

25 L

CAS Number:  112-80-1
Price
201,00 €
Log in for availability
1 * 25 L
Product Details & Documents

Synonyms: (Z)-9-Octadecenoic acid.

Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Boiling Pt:286 °C
  • Melting Pt:4 °C
  • Density:0.890 kg/l

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Assay (G.C.):
    65,0 - 88,0%
  • Appearance:
    Passes test
  • Residue on ignition (as SO₄):
    0,1%
  • Acidity value:
    195 - 204
  • Iodine value:
    89 - 105
  • Peroxide value:
    10,0
  • Residual solvents (Ph. Eur.):
    Passes test
  • Myristic acid:
    5,0%
  • Palmitic acid:
    16,0%
  • Palmitoleic acid:
    8,0%
  • Stearic acid:
    6,0%
  • Linoleic acid:
    18,0%
  • Linolenic acid:
    4,0%
  • Fatty acids with chain length greater than C18:
    4,0%
  • Margaric acid:
    4,0%
  • Identity:
    IR passes test
  • Cadmium (Cd) - Class 1 impurities (ICH Q3D):
    0,5 ppm
  • Lead (Pb) - Class 1 impurities (ICH Q3D):
    2 ppm
  • Arsenic (As) - Class 1 impurities (ICH Q3D):
    1,5 ppm
  • Mercury (Hg) - Class 1 impurities (ICH Q3D):
    1,5 ppm
  • Cobalt (Co) - Class 2A impurities (ICH Q3D):
    5 ppm
  • Vanadium (V) - Class 2A impurities (ICH Q3D):
    10 ppm
  • Nickel (Ni) - Class 2A impurities (ICH Q3D):
    20 ppm
  • Thallium (Tl) - Class 2B impurities (ICH Q3D):
    5 ppm
  • Gold (Au) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Palladium (Pd) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Iridium (Ir) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Osmium (Os) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Rhodium (Rh) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Ruthenium (Ru) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Selenium (Se) - Class 2B impurities (ICH Q3D):
    15 ppm
  • Silver (Ag) - Class 2B impurities (ICH Q3D):
    15 ppm
  • Platinum (Pt) - Class 2B impurities (ICH Q3D):
    10 ppm
  • Lithium (Li) - Class 3 impurities (ICH Q3D):
    55 ppm
  • Antimony (Sb) - Class 3 impurities (ICH Q3D):
    120 ppm
  • Barium (Ba) - Class 3 impurities (ICH Q3D):
    140 ppm
  • Molybdenum (Mo) - Class 3 impurities (ICH Q3D):
    25 ppm
  • Copper (Cu) - Class 3 impurities (ICH Q3D):
    250 ppm
  • Tin (Sn) - Class 3 impurities (ICH Q3D):
    600 ppm
  • Chromium (Cr) - Class 3 impurities (ICH Q3D):
    25 ppm

Specifications

  • Pk:
    25 L

Specifications