Orders LinkContactUsLinkComponent
L-Carnosine
Bottle
Catalog # GENORC-681
Supplier:   G-Biosciences
L-Carnosine
Catalog # GENORC-681
Supplier:  G-Biosciences
CAS Number:  
L-Carnosine
Bottle
CAS Number:  
Price
3 370,00 €
Log in for availability
1 * 500 g
Product Details & Documents
L-Carnosine is a white to slightly off-white crystalline powder that is used as a substrate for carnosinase-1 and other enzymes.
  • Molecular Biology Grade
Specifications
Specifications
  • Pack type:
    Bottle
  • Cat. No.:
    GENORC-681
  • Protein/peptide name:
    L-Carnosine
Specifications