G-Biosciences Acetate Buffer
Acetate buffer, pH 5,5 (200 mM), 250 ml
Catalog # GENO786-877
Supplier: G-Biosciences
Jump to:
Product Details & Documents
Acetate buffer at pH 5,0 and 5,5 are supplied as ready to use solutions that eliminate the calculation, formulation and weighing errors.
- Precision in pH
- Guaranteed reproducibility
Acetate buffer, pH 5,0 buffer is commonly used in protein purification, enzyme stabilisation, and histology.
Acetate buffer, pH 5,5 is useful for preserving enzyme activity or biomolecules that are sensitive to lower pH.
Specifications
Specifications
- Description:Acetate buffer, pH 5,5 (200 mM)
- Volume:250 ml