Order Entry
Export
Orders LinkContactUsLinkComponent
G-Biosciences Acetate Buffer

G-Biosciences Acetate Buffer

Acetate buffer, pH 5,0 (200 mM), 250 ml
Catalog # GENO786-876
Supplier:   G-Biosciences
G-Biosciences Acetate Buffer
Catalog # GENO786-876
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences Acetate Buffer

Acetate buffer, pH 5,0 (200 mM), 250 ml

CAS Number:  
Price
71,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Acetate buffer at pH 5,0 and 5,5 are supplied as ready to use solutions that eliminate the calculation, formulation and weighing errors.

  • Precision in pH
  • Guaranteed reproducibility

Acetate buffer, pH 5,0 buffer is commonly used in protein purification, enzyme stabilisation, and histology.

Acetate buffer, pH 5,5 is useful for preserving enzyme activity or biomolecules that are sensitive to lower pH.

Specifications

Specifications

  • Description:
    Acetate buffer, pH 5,0 (200 mM)
  • Volume:
    250 ml

Specifications