Order Entry
Export
Orders LinkContactUsLinkComponent
Hygromycin B

Hygromycin B

Bottle
Catalog # GENORC-178
Supplier:   G-Biosciences
Hygromycin B
Catalog # GENORC-178
Supplier:  G-Biosciences
CAS Number:  31282-04-9
Hygromycin B

Bottle

CAS Number:  31282-04-9
Price
1 360,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Mode of action and Anti-microbial spectrum: Hygromycin B is an aminocyclitol with a wide spectrum of activity that kills bacteria, fungi and higher eukaryotic cells by inhibiting protein synthesis. It is produced by the bacterium Streptomyces hygroscopicus.

  • Molecular Biology Grade
  • Tested against both sensitive and resistant cells
Synonyms:  (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
  • CAS Number:31282-04-9
  • Formula:C₂₀H₃₇N₃O₁₃
  • Molecular Weight:527,6 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications