Chromium standard solution, VHG
Chromium III, Cr, 100 mg/L, 2% HNO₃, 100 mL
Catalog # PROOVHG-SPCR3-100
Supplier: LGC Standards PROMOCHEM
Chromium standard solution, VHG
Catalog # PROOVHG-SPCR3-100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Jump to:
Product Details & Documents
This standard solution is intended for use as a calibration and reference standard for plasma emission spectroscopy (ICP or DCP), inductively coupled plasma mass spectroscopy (ICP-MS), atomic absorption spectroscopy (AA or GFAA) or techniques using other modes of elemental detection and an appropriate separation method.
Specifications
Specifications
- Concentration:100 mg/L
- Matrix:2% HNO₃
- Pk:100 mL
- Element:Chromium III
- Symbol:Cr



