Needles replacement parts
Needle seat, standard, PEEK, 0,17 mm ID, 1100, 1120, 1200, 1220, 1260, G1313A, G1329A/B, G1329-87017, AGLS
Catalog # CTSA11-2605
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Specifications
- Description:Needle seat, standard, PEEK, 0,17 mm ID
- Seq:11
- OEM manufacturer:AGLS
- OEM equivalent:G1329-87017
- Model:1100, 1120, 1200, 1220, 1260, G1313A, G1329A/B