Various replacement parts
Capillary assembly, 0,17ID×400L mm, with fittings, 1100, 1200, 1260, G1312, G1312-87303, AGLS
Catalog # CTSACTS-A11459
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
- Description:Capillary assembly, 0,17ID×400L mm, with fittings
- OEM manufacturer:AGLS
- OEM equivalent:G1312-87303
- Model:1100, 1200, 1260, G1312