Various replacement parts
Standard zirconia mixer, 50 µl, ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM, 700002911, WATE
Catalog # CTSACTS-A11135
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Standard zirconia mixer, 50 µl
- Seq:54
- OEM manufacturer:WATE
- OEM equivalent:700002911
- Model:ACQUITY UPLC I2V BSM, ACQUITY UPLC BSM