Orders LinkContactUsLinkComponent
Various replacement parts
Compression screw, short, 6,35 mm, gold-plated, Acquity H-Class Bio Quaternary Solvent Manager, Acquity I-Class BSM, Acquity UPLC Binary Solvent Manager, nanoAcquity UPLC Sample Manager, Open Architecture UPLC, 700002634, WATE
  CTSA11-2525
 :   SCIENCIX
Various replacement parts
  CTSA11-2525
 :  SCIENCIX
Various replacement parts
Various replacement parts
Compression screw, short, 6,35 mm, gold-plated, Acquity H-Class Bio Quaternary Solvent Manager, Acquity I-Class BSM, Acquity UPLC Binary Solvent Manager, nanoAcquity UPLC Sample Manager, Open Architecture UPLC, 700002634, WATE
173,00 €
1 * 10 items
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
  • Description:
    Compression screw, short, 6,35 mm, gold-plated
  • Seq:
    12
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    700002634
  • Model:
    Acquity H-Class Bio Quaternary Solvent Manager, Acquity I-Class BSM, Acquity UPLC Binary Solvent Manager, nanoAcquity UPLC Sample Manager, Open Architecture UPLC