Orders LinkContactUsLinkComponent
Various replacement parts
Various replacement parts
Capillary, 0,17ID×400L mm, 1100, 1200, 1260, 5021-1819, AGLS
Catalog # CTSACTS-11459
Supplier:   SCIENCIX
Various replacement parts
Catalog # CTSACTS-11459
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Capillary, 0,17ID×400L mm, 1100, 1200, 1260, 5021-1819, AGLS
Price
50,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    Capillary, 0,17ID×400L mm
  • Seq:
    09
  • OEM manufacturer:
    AGLS
  • OEM equivalent:
    5021-1819
  • Model:
    1100, 1200, 1260