Various replacement parts
Seal cap assembly, 1050, 1100, 1200, 1220, 1260, 5067-4728, AGLS
CTSACTS-A11454
: SCIENCIX
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
- Description:Seal cap assembly
- Seq:49
- OEM manufacturer:AGLS
- OEM equivalent:5067-4728
- Model:1050, 1100, 1200, 1220, 1260