Various replacement parts
Fingertight one piece 10-32 PEEK male nut/ferrule for 1/16" OD tubing, (Agilent) 1050, 1090, 1100, 1200, 1220
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000, 5063-6591
ED020005, Agilent
PerkinElmer
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000, 5063-6591
ED020005, Agilent
PerkinElmer
CTSA11-1240
: SCIENCIX
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
- Description:Fingertight one piece 10-32 PEEK male nut/ferrule for 1/16" OD tubing
- Seq:27
- OEM manufacturer:Agilent
PerkinElmer - OEM equivalent:5063-6591
ED020005 - Model:(Agilent) 1050, 1090, 1100, 1200, 1220
(PerkinElmer) 200 Series, 1,2,3,3B, 4,10, 250, 400, 410, 620, Int. 4000