Orders LinkContactUsLinkComponent
Various replacement parts
Ferrule set, 1,58 mm ID, two-piece, ACQUITY CM-30S, ACQUITY H-Class Bio QSM, ACQUITY H-Class QSM, ACQUITY H-Class Bio SM-FTN, ACQUITY I-Class BSM, ACQUITY UPLC BSM, Open Architecture UPLC, nanoACQUITY HDX Mgr, nanoACQUITY UPLC BSM/ASM, 700002635, WATE
Catalog # CTSA11-2526
Supplier:   SCIENCIX
Various replacement parts
Catalog # CTSA11-2526
Supplier:  SCIENCIX
Various replacement parts
Various replacement parts
Ferrule set, 1,58 mm ID, two-piece, ACQUITY CM-30S, ACQUITY H-Class Bio QSM, ACQUITY H-Class QSM, ACQUITY H-Class Bio SM-FTN, ACQUITY I-Class BSM, ACQUITY UPLC BSM, Open Architecture UPLC, nanoACQUITY HDX Mgr, nanoACQUITY UPLC BSM/ASM, 700002635, WATE
Price
95,10 €
Log in for availability
1 * 10 items
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Specifications
  • Description:
    Ferrule set, 1,58 mm ID, two-piece
  • Seq:
    18
  • OEM manufacturer:
    WATE
  • OEM equivalent:
    700002635
  • Model:
    ACQUITY CM-30S, ACQUITY H-Class Bio QSM, ACQUITY H-Class QSM, ACQUITY H-Class Bio SM-FTN, ACQUITY I-Class BSM, ACQUITY UPLC BSM, Open Architecture UPLC, nanoACQUITY HDX Mgr, nanoACQUITY UPLC BSM/ASM