Various replacement parts
Line filter assembly, i-Series, LC-10ADvp, LC-10AT, LC-10ATvp, LC-20AD/AB, LC-20ADXR, LC-20AR, LC-20AT, LC-30ADSF, LC-2010, 228-35871-96, SHIM
Catalog # CTSACTS-A11421
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Line filter assembly
- OEM manufacturer:SHIM
- OEM equivalent:228-35871-96
- Model:i-Series, LC-10ADvp, LC-10AT, LC-10ATvp, LC-20AD/AB, LC-20ADXR, LC-20AR, LC-20AT, LC-30ADSF, LC-2010