Various replacement parts
Diaphragm, PTFE, LC-10AD, LC-10ADvp, LC-20AD/AB, LC-20ADXR, LC-30ADSF, LC-2010, LC-2010 HT, 228-32784-91, SHIM
Catalog # CTSA11-2891
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Diaphragm, PTFE
- OEM manufacturer:SHIM
- OEM equivalent:228-32784-91
- Model:LC-10AD, LC-10ADvp, LC-20AD/AB, LC-20ADXR, LC-30ADSF, LC-2010, LC-2010 HT