Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Product Details & Documents
Eppendorf Tubes® with snap caps are an ideal choice when working with medium-sized sample volumes (0,5 to 5,0 ml). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25000×g), and sample recovery.
- Designed for ease of use and user-friendly handling during sample preparation and processing
- Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions
- Specifically designed to enhance sample recovery, preserving the integrity of samples
- Available in Eppendorf Quality, PCR Clean, Biopur®, Sterile, and Forensic DNA Grade quality for tested purity
Explore the versatility of Eppendorf 5,0 ml tubes, available in both colorless and amber options. Tailor the selection with various certified purities, ensuring precise and reliable sample recoveries. Experience the flexibility these tubes provide, allowing selection of the ideal combination for specific research needs.
Manufactured without the use of slip agents, plasticisers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Cap type: Snap cap
Material: PP
Diameter: 17 mm
Certifications: Certificate of Quality/Conformity, Lot-specific certificates for the purity grades PCR clean, Biopur, Sterile®, Forensic DNA Grade can be downloaded from the Eppendorf website.
Specifications
- Packed:200 tubes (10 bags × 20 tubes)
- Capacity:5,0 ml
- Colour:Colourless
- Cat. No.:525-0797
- Operating temperature range:−86...100 °C
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Volume:5,0 ml
- Max. RCF:25000×g
- Autoclavable:When open 121 °C, 20 min
- Environmentally Preferable:Low Manufacturing Impact



