Orders LinkContactUsLinkComponent
Reactive quencher dyes
Reactive quencher dyes
CFQ520 maleimide, 5 mg
Catalog # BTIU80502
Supplier:   Biotium
Reactive quencher dyes
Catalog # BTIU80502
Supplier:  Biotium
CAS Number:  
Reactive quencher dyes
Reactive quencher dyes
CFQ520 maleimide, 5 mg
CAS Number:  
Price
435,00 €
Log in for availability
1 * 5 mg
Product Details & Documents
Reactive quenchers that act as acceptors in fluorescence resonance energy transfer (FRET) applications.
  • Thiol- and amine-reactive quenchers
  • One Sample Fluorescence Resonance Energy Transfer (OS-FRET) and FRET quenchers
NFQ1, a non-fluorescent quencher, is used for the One Sample Fluorescence Resonance Energy Transfer (OS-FRET) method employing a non-fluorescent methanethiosulfonate-linked acceptor that can be reversibly coupled to a target sulfhydryl residue via a disulfide bond. After the quenched donor emission is quantitated the acceptor is removed by reduction, enabling measurement of unquenched donor emission in the same sample.
The thiol-reactive quencher, CFQ520 maleimide, has a broad and intense absorption (approximately 520 nm) but no fluorescence, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
The amine-reactive quencher, CFQ520 succinimidyl ester, has a broad and intense absorption (approximately 520 nm) but no fluorescence, making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.
N-(6-Aminohexyl)maleimide is a cross-linking reagent that can react with an amine-reactive group and a thiol group.
Specifications
Specifications
  • Pk:
    5 mg
  • Description:
    CFQ520 maleimide
Specifications