0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Buffer Solution pH 7.02 (20°C) 1 * 1.000 mL
APPC273108.1211
:
PanReac AppliChem
Buffer Solution pH 7.02 (20°C) 1 * 1.000 mL
APPC273108.1211
:
PanReac AppliChem