Orders LinkContactUsLinkComponent
Labels, hazard warning, electricity
Hazard labels, PIC 307-TRI 025-B7541, Polyester, Black on yellow, 25×22 mm, 13 labels/card
  116-0681
 :   Brady
Labels, hazard warning, electricity
  116-0681
 :  Brady
Labels, hazard warning, electricity
Labels, hazard warning, electricity
Hazard labels, PIC 307-TRI 025-B7541, Polyester, Black on yellow, 25×22 mm, 13 labels/card
9,30 €
1 * 1 items
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
  • Self-adhesive
  • Clear identification of voltage, phases, grounding and insulation
  • Colour:
    Black on yellow
  • Description:
    Hazard labels
  • Dimensions:
    25×22 mm
  • Material:
    Polyester
  • Seq:
    01
  • Model:
    PIC 307-TRI 025-B7541
  • Packed:
    13 labels/card