Sodium hydroxide solution 40% (w/w)
25 L
Catalog # APPC171220.0716
Supplier: PanReac AppliChem
Sodium hydroxide solution 40% (w/w)
Catalog # APPC171220.0716
Supplier: PanReac AppliChem
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Absorbs CO₂ from air. Miscible with water.
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Density:1.43 g/ml
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Minimum Assay (Acidim.):40%
- Nitrogen Compounds (as N):Max. 0.002%
- Carbonate (as Na₂CO₃):Max. 1.0%
Specifications
- Pk:25 L



