Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
L(+)-Ascorbic Acid for analysis, ACS 1 * 100 g
L(+)-Ascorbic Acid for analysis, ACS 1 * 100 g
Print
L(+)-Ascorbic Acid for analysis, ACS 1 * 100 g
Catalog # APPC131013.1208
Certificates
Supplier:
PanReac AppliChem
L(+)-Ascorbic Acid for analysis, ACS 1 * 100 g
Catalog # APPC131013.1208
Supplier:
PanReac AppliChem
CAS Number:
Certificates
Expand image
Return to Top
L(+)-Ascorbic Acid for analysis, ACS 1 * 100 g
Add to cart
CAS Number:
Price
51,80 €
Log in
for availability
Unit of Measure
1 * 100 g
-
+
Add to cart
Return Policy
Product Details & Documents
Documents
Certificates
Search by Lot Number
Product specification
APPC131013.1208_TDS_EN