Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 10 L
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 10 L
Print
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 10 L
Catalog # APPC273617.1315
Supplier:
PanReac AppliChem
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 10 L
Catalog # APPC273617.1315
Supplier:
PanReac AppliChem
Expand image
Return to Top
Buffer Solution pH 7.00 (20°C) (yellow colour) 1 * 10 L
Add to cart
Price
228,00 €
Log in
for availability
Unit of Measure
1 * 10 L
-
+
Add to cart
Return Policy
Product Details & Documents