Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium hypochlorite solution 13% (w/v), Technical Grade

Sodium hypochlorite solution 13% (w/v), Technical Grade

10 L
Catalog # APPC213322.0715
Corrosive
Environment
Supplier:   PanReac AppliChem
Sodium hypochlorite solution 13% (w/v), Technical Grade
Catalog # APPC213322.0715
Supplier:  PanReac AppliChem
CAS Number:  7681-52-9
Sodium hypochlorite solution 13% (w/v), Technical Grade

10 L

CAS Number:  7681-52-9
Price
63,90 €
Log in for availability
1 * 10 L
Product Details & Documents
  • CAS Number:7681-52-9
  • Formula:ClNaO
  • MDL Number:MFCD00011120
  • Molecular Weight:74.44 g/mol
  • Storage Temperature:Ambient
  • UN:1791
  • ADR:8,III
Specifications

Specification Test Results

  • Assay (Active Chlorine, Iodometry):
    10.0 - 14.0%
  • NaCl:
    Max. 11.5%

Specifications

  • Pk:
    10 L

Specifications