Sodium hypochlorite solution 13% (w/v), Technical Grade
5 L
Catalog # APPC213322.1214
Supplier: PanReac AppliChem
Sodium hypochlorite solution 13% (w/v), Technical Grade
Catalog # APPC213322.1214
Supplier: PanReac AppliChem
CAS Number: 7681-52-9
Jump to:
Product Details & Documents
- CAS Number:7681-52-9
- Formula:ClNaO
- MDL Number:MFCD00011120
- Molecular Weight:74.44 g/mol
- Storage Temperature:Ambient
- UN:1791
- ADR:8,III
Specifications
Specification Test Results
- Assay (Active Chlorine, Iodometry):10.0 - 14.0%
- NaCl:Max. 11.5%
Specifications
- Pk:5 L



