Venipuncture sets, Surflo®
Winged infusion sets with protector, G 23, Light blue, 19 mm
TERUSV-S23NL30
: TERUMO
Surshield prevents needle stick injuries with painless placement.
Cannula sharpness combined with a special silicone coating allows an easy and smooth placement.
- Gauge:G 23
- Colour:Light blue
- Description:Winged infusion sets with protector
- Length:19 mm
- Seq:003