Orders LinkContactUsLinkComponent
Multi-parameter meters, handheld, Portavo® 907
Portavo® 907 LDO Set, with optical dissolved oxygen sensor SE340-MS and carrying case
Catalog # 665-0361
Supplier:   KNICK
Multi-parameter meters, handheld, Portavo® 907
Catalog # 665-0361
Supplier:  KNICK
Multi-parameter meters, handheld, Portavo® 907
Multi-parameter meters, handheld, Portavo® 907
Portavo® 907 LDO Set, with optical dissolved oxygen sensor SE340-MS and carrying case
Price
2 722,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
These versatile, portable instruments can measure pH, ORP, conductivity and dissolved oxygen (optical). Thanks to their robustness and large selection of suitable sensors, they are suitable for both laboratory and process applications. Waterproof according to IP 66/IP 67.
  • Reinforced plastic enclosure for rough industrial applications
  • Integrated cover protects the display and serves as laboratory stand
  • High resolution colour TFT display is transreflective allowing the display to be read in bright sunlight
  • Data log function can store up to 10000 values
  • Digital Memosens® input, which allows signals to be digitised and transferred inductively i.e. via a non-contact connection
  • Integrated sensor quiver (protective container for sensor) prevents sensor dry-out and damage
  • Optical oxygen measurements possible
  • Automatic status check of the device and sensor; evaluation of the sensor data end calibration intervals
  • Memosens®: All information on sensor life are saved directly on the sensor (history of calibration, operation time, lifetime, sensor wear, SIP cycles…)
Portavo® 907 OXY Set includes the Portavo® 907 multi-parameter-meter and the digital probe SE 340 for optical oxygen measurements, with M12 8-pin plug according to DIN EN 50262.
Portavo® 907 LDO Set includes the Portavo® 907 multi-parameter-meter, the digital probe SE 340 for optical oxygen measurements, a carrying case and buffer solutions.
Portavo® 907 pH Set includes the Portavo® 907 multi-parameter-meter, the digital Memosens sensor SE 102 NMS, Memosens cable, carrying case and pH buffer solutions.
Portavo® 907 Multi Set includes the Portavo® 907 multi-parameter-meter, the digital Memosens pH sensor SE 102 NMS, Meosens conductivity sensor SE 615/1-MS, Memosens oxygen sensor SE 715/1-MS, Memosens cable, carrying case and pH buffer solutions.
 : Supplied with manual, 4× AA batteries, carrying strap, factory certificate, micro USB port for PC connection, USB cable (1,5 m) and Paraly® 112 PC software. All instruments, except for the sets, are delivered without an electrode, which needs to be ordered separately.
Portavo® 907 LDO Set includes the Portavo® 907 multi-parameter meter, SE340-MS digital probe for optical oxygen measurements, a carrying case and buffer solutions.
Portavo® 907 pH Set includes the Portavo® 907 multi-parameter meter, Memosens SE102-MS digital pH sensor, Memosens cable CA/MS-001XFA-L, carrying case and pH buffer solutions.
Portavo® 907 Multi Set includes the Portavo® 907 multi-parameter meter, Memosens SE102-MS digital pH sensor, Memosens SE615/1-MS conductivity sensor, Memosens SE715/1-MS oxygen sensor, Memosens cable CA/MS-001XFA-L, carrying case and pH buffer solutions.
Specifications
  • Description:
    Portavo® 907 LDO Set, with optical dissolved oxygen sensor SE340-MS and carrying case
  • Seq:
    05
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • pH buffer set CaliMat, 4,00/7,00/9,00, ready-to-use; 250 ml each
    pH buffer set CaliMat, 4,00/7,00/9,00, ready-to-use; 250 ml each
    662-1826
    Log in for availability
    1 * 1 SET - 45,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PT1000 temperature probe, –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751
    PT1000 temperature probe, –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751
    662-3532
    Log in for availability
    1 * 1 items - 257,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Adapter for connecting conductivity sensors with 2 banana plugs with instrument
    Adapter for connecting conductivity sensors with 2 banana plugs with instrument
    665-0300
    Log in for availability
    1 * 1 items - 109,00 €
Product Information
  • Catalog #
    Availability
    Price
  • KPG® tube for 4-electrode conductivity sensor ZU 6985, incl. O-ring
    KPG® tube for 4-electrode conductivity sensor ZU 6985, incl. O-ring
    665-0298
    Log in for availability
    1 * 1 items - 85,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Adapter for connecting 4 electrode sensor ZU 6985 with instrument
    Adapter for connecting 4 electrode sensor ZU 6985 with instrument
    665-0301
    Log in for availability
    1 * 1 items - 109,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Robust field case (for meter, sensor, various small parts and user manual)
    Robust field case (for meter, sensor, various small parts and user manual)
    662-1599
    Log in for availability
    1 * 1 items - 154,00 €
Product Information
  • Catalog #
    Availability
    Price
  • PT1000 temperature probe, with tilted tip for puncture measurements in semi-solid substances; –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751
    PT1000 temperature probe, with tilted tip for puncture measurements in semi-solid substances; –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751
    663-2215
    Log in for availability
    1 * 1 items - 257,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Conductivity standard, 12,88 mS/cm ±1 %, one ampoule for producing 1000 ml 0,1 mol/l NaCl solution
    Conductivity standard, 12,88 mS/cm ±1 %, one ampoule for producing 1000 ml 0,1 mol/l NaCl solution
    663-2112
    Log in for availability
    1 * 1 items - 57,00 €