Order Entry
Export
Orders LinkContactUsLinkComponent
Accessories for test sieves

Accessories for test sieves

Intermediate pan, 200 mm, 50 mm
  HAVE205923715
 :   NEXOPART
Accessories for test sieves
  HAVE205923715
 :  NEXOPART
Accessories for test sieves
Accessories for test sieves

Intermediate pan, 200 mm, 50 mm

 
92,90 €
   
1 * 1 items
 
 
  • Ø:
    200 mm
  • Description:
    Intermediate pan
  • Height:
    50 mm
  • Seq:
    02