Order Entry
Export
Orders LinkContactUsLinkComponent
 

G418 disulphate solution 50 mg/ml

Plastic bottle, 100 ml, 1
  MDTC30-234-CI
Health
 :   Corning
G418 disulphate solution 50 mg/ml
  MDTC30-234-CI
 :  Corning
 :  108321-42-2
G418 disulphate solution 50 mg/ml
G418 disulphate solution 50 mg/ml

Plastic bottle, 100 ml, 1

 :  108321-42-2
 
565,98 €
Log in for availability
1 * 1 items
 

G418 Sulfate is recommended for use as selection agent in a concentration range of 100-800 µg/ml.

This antibiotic blocks polypeptide synthesis by hindering protein elongation, and inhibits the selection and maintenance of eukaryotic cells stably transfected with the neomycin resistance gene. G418 is an aminoglycoside similar in structure to gentamicin, neomycin and kanamycin.

 : For Research or Further Manufacturing Use Only. Not for use in Diagnostic Procedures.

Synonyms:  G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
  • CAS Number:108321-42-2
  • Formula:C₂₀H₄₄N₄O₁₈S₂
  • MDL Number:MFCD00058314
  • Molecular Weight:692.72 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1
  • Pk Info:
    100 ml

Specifications