ContactUsLinkComponent
Double-pass spray chamber, inert, PTFE cyclonic design with ball joint socket and UniFit drain outlet, for use with HF digests and h igh TDS samples with Agilent 5100 ICP-OES 1 * 1 items
Catalog # HEWLG8010-80034
Supplier:  Agilent
undefined
Double-pass spray chamber, inert, PTFE cyclonic design with ball joint socket and UniFit drain outlet, for use with HF digests and h igh TDS samples with Agilent 5100 ICP-OES 1 * 1 items
Catalog # HEWLG8010-80034
Supplier:  Agilent