Order Entry
Export
Orders LinkContactUsLinkComponent
 

Multi-Layer Foil Gas Sampling Bags

Gas sampling bags, 3 L, 254×254 mm
Catalog # REST22951
Supplier:   Restek
Multi-Layer Foil Gas Sampling Bags
Catalog # REST22951
Supplier:  Restek
Multi-Layer Foil Gas Sampling Bags
Multi-Layer Foil Gas Sampling Bags

Gas sampling bags, 3 L, 254×254 mm

Price
116,00 €
Log in for availability
1 * 5 items
Product Details & Documents

Sampling bags are cost-effective air sampling device for high level VOC and permanent gases. It features a PP combo valve with hose connection to fit 4,76 mm ID tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.

  • Good stability for low molecular weight compounds, such as methane, CO, CO₂ and permanent gases
  • Chemically inert with light and moisture protection
  • Not recommended for low ppm VOC due to background levels
  • Protective five layer barrier minimises gas permeability
  • Continuous sampling temperature up to 88 °C
Specifications
  • Capacity:
    3 L
  • Description:
    Gas sampling bags
  • Seq:
    02
  • W×L:
    254×254 mm