Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POLYETHYLENE STOPPERS WITH OCTAGONAL PLATE, ST 34/35, ACC. TO DIN 12254 1 * 1 items
Catalog # WITG0.306.034
Supplier:
witeg Labortechnik
Expand image
POLYETHYLENE STOPPERS WITH OCTAGONAL PLATE, ST 34/35, ACC. TO DIN 12254 1 * 1 items
Catalog # WITG0.306.034
Supplier:
witeg Labortechnik