Order Entry
Export
Orders LinkContactUsLinkComponent
Septa caps

Septa caps

Open top septum cap, bonded 1,5 mm, PTFE-lined silicon septa, 24-414
  EPSC297-2400UB
   Low Manufacturing Impact
Septa caps
  EPSC297-2400UB
   Low Manufacturing Impact
Septa caps
Septa caps

Open top septum cap, bonded 1,5 mm, PTFE-lined silicon septa, 24-414

 
77,50 €
Log in for availability
1 * 24 items
 

PP, white.

Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.

Specifications
  • Thread:
    24-414
  • Description:
    Open top septum cap, bonded 1,5 mm, PTFE-lined silicon septa
  • Seq:
    04
  • Cat. No.:
    EPSC297-2400UB
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories

Product Information

  •  
    Type
    For
    Ø
     
     
  • EPSC288-0022
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 24 items - 64,80 €

Product Information

  •  
    Type
    For
    Ø
     
     
  • EPSC288-7222
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 72 items - 157,00 €

Product Information

  •  
    Type
    For
    Ø
     
     
  • EPSC288-0022BP
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 2.000 items - 2 430,00 €

Product Information

  •  
    Type
    For
    Ø
     
     
  • EPSCS24-400-S2
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 144 items - 287,00 €