Orders LinkContactUsLinkComponent
Septa caps
Septa caps
Open top cap, without septum, 24-414
Catalog # EPSC297-2414
   Low Manufacturing Impact
Supplier:   EP SCIENTIFIC
Septa caps
Catalog # EPSC297-2414
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Septa caps
Septa caps
Open top cap, without septum, 24-414
Price
64,70 €
Log in for availability
1 * 24 items
Product Details & Documents
PP, white.
Septa caps fit VOA vials, bottles and jars. The septum is permanently bonded to the cap and processed to meet EPA performance based specifications for volatile organic analysis.
Specifications
  • Thread:
    24-414
  • Description:
    Open top cap, without septum
  • Seq:
    05
  • Cat. No.:
    EPSC297-2414
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Type
    For
    Ø
    Availability
    Price
  • EPSC288-0022
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 24 items - 64,80 €
Product Information
  • Catalog #
    Type
    For
    Ø
    Availability
    Price
  • EPSC288-7222
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 72 items - 157,00 €
Product Information
  • Catalog #
    Type
    For
    Ø
    Availability
    Price
  • EPSC288-0022BP
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 2.000 items - 2 430,00 €
Product Information
  • Catalog #
    Type
    For
    Ø
    Availability
    Price
  • EPSCS24-400-S2
    Silicone, PTFE-lined
    24-214 VOA vials
    24 mm
    Log in for availability
    1 * 144 items - 287,00 €