Volumetric flasks, class A, blue markings, BLAUBRAND®
10 ml, 10/19, 0,04 ± ml, Wide neck, with PP stopper
Catalog # BRND2.37243
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, supplied with batch certificate
* In addition to DIN.
Certifications: DIN EN ISO 1042.
Documents
Certificates
Search by Lot NumberBrand Volumetric Measurement in the Laboratory
Product Literature
BRND2.37290_TDS_EN
Datasheet
Specifications
- NS:10/19
- Capacity:10 ml
- Description:Wide neck, with PP stopper
- Seq:02
- Tolerance:0,04 ± ml