Volumetric flasks, class A, blue markings, BLAUBRAND®
50 ml, 12/21, 0,06 ± ml, Narrow mouth, with PP stopper
Catalog # BRND2.37248
Supplier: Brand
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, supplied with batch certificate
* In addition to DIN.
: DIN EN ISO 1042.
Documents
Certificates
Search by Lot NumberBrand Volumetric Measurement in the Laboratory
Product Literature
BRND2.37290_TDS_EN
Datasheet
Specifications
- NS:12/21
- Capacity:50 ml
- Description:Narrow mouth, with PP stopper
- Seq:06
- Tolerance:0,06 ± ml