Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MATRIX METALLOPROTEINASE 11 MM MAB MO XH 1 * 250 µl
Catalog # USBIM2427-29-250
Supplier:
US Biological
Expand image
MATRIX METALLOPROTEINASE 11 MM MAB MO XH 1 * 250 µl
Catalog # USBIM2427-29-250
Supplier:
US Biological
Related Information
Documentation
About USBiological
Documentation