Orders LinkContactUsLinkComponent
Galtek® check valves
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm
Catalog # ENTECK4-4N-4N
Supplier:   ENTEGRIS
Galtek® check valves
Catalog # ENTECK4-4N-4N
Supplier:  ENTEGRIS
Galtek® check valves
Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
O-ring free design, ¹/₂ or ¹/₄" orifice.
  • O-ring free design reduces contamination
  • All wetted parts are molded PFA and PTFE for corrosion resistance
  • No metal parts are used that could corrode or contribute to contamination
  • Poppet design allows for a high flow through rate and maximises rinsability
Specifications
  • Description:
    Inlet 1/4" FNPT, outlet 1/4" FNPT, length 60,7 mm