Order Entry
Export
Orders LinkContactUsLinkComponent
Spray gun

Spray gun

PFA tubing, coiled, O.Ø 9,5 mm, with 3,2 mm recirculation tubing, preassembled
Catalog # ENTE221-69
Supplier:   ENTEGRIS
Spray gun
Catalog # ENTE221-69
Supplier:  ENTEGRIS
Spray gun

PFA tubing, coiled, O.Ø 9,5 mm, with 3,2 mm recirculation tubing, preassembled

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

High purity PFA (all wetted parts).

  • With 4,8 mm orifice and ³/₈" Flaretek® tube fitting or ¹/₂" compression connections
  • Max. pressure 5,52 bar

Used in fume hoods and wet processing stations to help protect against corrosion.

Specifications
  • Description:
    PFA tubing, coiled, O.Ø 9,5 mm, with 3,2 mm recirculation tubing, preassembled
  • Seq:
    11
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Repair tool to access front and rear diaphragm of gun
    Repair tool to access front and rear diaphragm of gun
    ENTE213-80
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
    ENTE421-06
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap, natural
    Replacement cap, natural
    ENTE600037
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement nozzle
    Replacement nozzle
    ENTE500121
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap, orange
    Replacement cap, orange
    ENTE600037OR
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Availability
    Price
  • Replacement cap, blue
    Replacement cap, blue
    ENTE600037BL
    Log in for availability
    1 * 1 items