Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Perforated basket, O 370 mm, height 250 mm,for Autoclaves WAC-80 1 * 1 items
Catalog # WITGDH.WAC20080
Supplier:
witeg Labortechnik
Expand image
Perforated basket, O 370 mm, height 250 mm,for Autoclaves WAC-80 1 * 1 items
Catalog # WITGDH.WAC20080
Supplier:
witeg Labortechnik