Order Entry
Export
Orders LinkContactUsLinkComponent
Digital dry block heaters

Digital dry block heaters

Dry Block Heater 1, 1, 165 W, RT*...120 °C, ±0,2 °C, 151×228×73 mm, 1 kg
Catalog # 460-0248
Digital dry block heaters
Catalog # 460-0248
Supplier:  
Digital dry block heaters
Digital dry block heaters

Dry Block Heater 1, 1, 165 W, RT*...120 °C, ±0,2 °C, 151×228×73 mm, 1 kg

Discontinued
Product Details & Documents

Digital block heaters which hold from one up to four aluminum blocks, provide precise temperature control in small vessels. The PT1000 temperature sensor allows temperature control directly in the sample vessel. It is used with PCR tubes, PCR strips, Greiner tubes, microplates and cuvettes. The block heaters are ideal for melting and boiling point determination, enzyme reactions, incubation and activation of cultures, immunoassays, DNA denaturation, culture media tests, coagulation tests, blood/urea nitrogen determinations and in situ hybridisations.

  • Stepless adjustable temperature from ambient up to 120 °C
  • Timer: Countdown, adjustable from 1 min to 99h 59 min
  • Counter: Shows heating time
  • Highly versatile with a large number of aluminium alloy blocks
  • Fixed safety circuit and error code display
  • Acoustic signal at end of test

Delivery information: Supplied with PT1000 temperature sensor, but without heating blocks, blocks must be ordered separately.

Caution: * RT = Ambient

Specifications
  • Heating capacity:
    165 W
  • No. of blocks:
    1
  • Temperature Range:
    RT*...120 °C
  • Weight:
    1 kg
  • Seq:
    02
  • W×D×H:
    151×228×73 mm
  • Temp. stability:
    ±0,2 °C
  • Type:
    Dry Block Heater 1
Accessories

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×11,5 mm Ø holes
    Single block, 20×11,5 mm Ø holes
    460-1009
    Eppendorf Tubes®, 1,5 ml
    36,9 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    Type
    Range
    Availability
    Price
  • Temperature sensor, coated (SafeCoat), DIN 45322
    Temperature sensor, coated (SafeCoat), DIN 45322
    460-1041
    PT 1000.90
    −10...+260 °C
    Log in for availability
    1 * 1 items - 239,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 80× 6,4 mm Ø strips
    Single block, 80× 6,4 mm Ø strips
    460-1036
    PCR tube strips (10×8 strips)
    15,5 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 30×7,9 mm Ø holes
    Single block, 30×7,9 mm Ø holes
    460-1008
    PCR tubes, 0,5 ml
    27,6 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 4×35 mm Ø holes
    Single block, 4×35 mm Ø holes
    460-1025
    Tubes, 35 mm, with round bottom
    47,6 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 12×17,5 mm Ø holes
    Single block, 12×17,5 mm Ø holes
    460-1022
    Tubes, 15 - 16 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Double block
    Double block
    460-1038
    96-well PCR plate (1)
    15,5 mm
    Log in for availability
    1 * 1 items - 338,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 30×8,3 mm Ø holes
    Single block, 30×8,3 mm Ø holes
    460-1018
    Tubes, 6 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 8×23,8 mm Ø holes
    Single block, 8×23,8 mm Ø holes
    460-1032
    Vials, 23 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 16×13,9 mm Ø holes
    Single block, 16×13,9 mm Ø holes
    460-1020
    Tubes, 12 - 13 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread
    Support rod, stainless steel, length 450 mm, Ø 10 mm, for all stirrers with M10 thread
    442-2041
    Log in for availability
    1 * 1 items - 88,50 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 12×17,8 mm Ø holes
    Single block, 12×17,8 mm Ø holes
    460-1029
    Vials, 17 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 5×29 mm Ø holes
    Single block, 5×29 mm Ø holes
    460-1014
    Conical tubes, 50 ml
    47,6 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Double block
    Double block
    460-1040
    96- or 384-well plate (1)
    13,5 mm
    Log in for availability
    1 * 1 items - 214,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, combination: 4/3/2×1,5/15/50 ml
    Single block, combination: 4/3/2×1,5/15/50 ml
    460-1016
    Centrifuge tubes, 1,5 ml/15 ml/50 ml
    39,1/44,5/47,6 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×15,8 mm Ø holes
    Single block, 20×15,8 mm Ø holes
    460-1028
    Vials, 15 mm
    35 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 6×28,8 mm Ø holes
    Single block, 6×28,8 mm Ø holes
    460-1034
    Vials, 28 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 8×25,8 mm Ø holes
    Single block, 8×25,8 mm Ø holes
    460-1033
    Vials, 25 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×13,9 mm Ø holes
    Single block, 20×13,9 mm Ø holes
    460-1021
    Tubes, 12 - 13 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 24×10,7 mm Ø holes
    Single block, 24×10,7 mm Ø holes
    460-1019
    Tubes, 10 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×10,9 mm Ø holes
    Single block, 20×10,9 mm Ø holes
    460-1012
    Corning tubes, 2,0 ml
    38,1 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×11,5 mm Ø holes
    Single block, 20×11,5 mm Ø holes
    460-1011
    VWR/Eppendorf Tubes®, 2,0 ml
    38,1 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 2×6 cuvettes 12,5 mm Ø
    Single block, 2×6 cuvettes 12,5 mm Ø
    460-1039
    Cuvettes
    25,4 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 8×21 mm Ø holes
    Single block, 8×21 mm Ø holes
    460-1023
    Tubes, 20 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Bosshead clamp
    Bosshead clamp
    442-2043
    Log in for availability
    1 * 1 items - 42,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 12×19,1 mm Ø holes
    Single block, 12×19,1 mm Ø holes
    460-1026
    Tubes, 17 - 18 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, combination: 6/10/5×0,5/1,5/2 ml
    Single block, combination: 6/10/5×0,5/1,5/2 ml
    460-1017
    Microtubes, 0,5 ml/1,5 ml/2,0 ml
    27,6/39,1/38,1 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 12×19,7 mm Ø holes
    Single block, 12×19,7 mm Ø holes
    460-1030
    Vials, 19 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×12,7 mm Ø holes
    Single block, 20×12,7 mm Ø holes
    460-1027
    Vials, 12 mm
    30 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 12×17,1 mm Ø holes
    Single block, 12×17,1 mm Ø holes
    460-1013
    Conical tubes, 15 ml
    44,5 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 6×26,2 mm Ø holes
    Single block, 6×26,2 mm Ø holes
    460-1024
    Tubes, 25 mm, with round bottom
    48,4 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 64× 6,4 mm Ø holes
    Single block, 64× 6,4 mm Ø holes
    460-1037
    PCR tubes (64)
    20,2 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Holding rod, for fastening ETS-D5 or VT-5 to the support rod
    Holding rod, for fastening ETS-D5 or VT-5 to the support rod
    431-2888
    Log in for availability
    1 * 1 items - 39,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 9×21,7 mm Ø holes
    Single block, 9×21,7 mm Ø holes
    460-1031
    Vials, 21 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, combination: 6/5/3×6/12/13/25 mm Ø holes
    Single block, combination: 6/5/3×6/12/13/25 mm Ø holes
    460-1015
    Test tubes, 6 ml/12 - 13 ml/25 ml
    48,4/48,4/48,4 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 20×11,1 mm Ø holes
    Single block, 20×11,1 mm Ø holes
    460-1010
    Microtubes 1,5 ml
    39,1 mm
    Log in for availability
    1 * 1 items - 140,00 €

Product Information

  • Catalog #
    For
    Depth
    Availability
    Price
  • Single block, 15×16,4 mm Ø holes
    Single block, 15×16,4 mm Ø holes
    460-1035
    Vials, 16 mm
    45 mm
    Log in for availability
    1 * 1 items - 147,00 €

Product Information

  • Catalog #
    Type
    Range
    Availability
    Price
  • Stainless steel temperature sensor, DIN 45322
    Stainless steel temperature sensor, DIN 45322
    442-0584
    PT 1000.60
    −20...+350 °C
    Log in for availability
    1 * 1 items - 169,00 €