Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
COLOR BUTTON RED PICUS 1 * 1 items
COLOR BUTTON RED PICUS 1 * 1 items
Print
COLOR BUTTON RED PICUS 1 * 1 items
Catalog # BHIT736026
Supplier:
Sartorius
COLOR BUTTON RED PICUS 1 * 1 items
Catalog # BHIT736026
Supplier:
Sartorius
Expand image
Return to Top
COLOR BUTTON RED PICUS 1 * 1 items
Add to cart
Price
68,40 €
Log in
for availability
Unit of Measure
1 * 1 items
-
+
Add to cart
Return Policy
Product Details & Documents