Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Cap Inserts
Inserts for Brand® cryogenic tubes
Inserts for Brand® cryogenic tubes
Print
Inserts for Brand® cryogenic tubes
White
Catalog # BRND114850
Supplier:
Brand
Inserts for Brand® cryogenic tubes
Catalog # BRND114850
Supplier:
Brand
Expand image
Return to Top
Inserts for Brand® cryogenic tubes
White
Specifications
Add to cart
Jump to:
Specifications
Price
17,80 €
Log in
for availability
Unit of Measure
1 * 500 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
PP inserts. Fit for all sizes.
Specifications
Colour:
White
Seq:
04