Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
POLARIMETER ADP 660 DIGITAL (UK PLUG) MULTIPLE WAVELENGTH 325,365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0791
Supplier:
BELLINGHAM STANLEY
Expand image
POLARIMETER ADP 660 DIGITAL (UK PLUG) MULTIPLE WAVELENGTH 325,365,405,436,546 & 589NM 1 * 1 items
Catalog # 635-0791
Supplier:
BELLINGHAM STANLEY
Related Information
Documentation
Videos
21 CFR part 11 Complaince using Polarimeters and Refractometers from Bellingham Sta
Touchscreen technology from Bellingham Stanley, a Xylem brand
Manual
Manual Digital Polarimetre ADP600 Series (Bellingham + Stanley)
Documentation