Order Entry
Export
Orders LinkContactUsLinkComponent
 

Bottles, narrow neck, with thread

150 ml
Catalog # SPVAC00748188
Supplier:   GRAVIS
Bottles, narrow neck, with thread
Catalog # SPVAC00748188
Supplier:  GRAVIS
Bottles, narrow neck, with thread
Bottles, narrow neck, with thread

150 ml

Price
108,00 €
Log in for availability
1 * 88 items
Product Details & Documents

Glass, clear, amber or yellow.

  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances

Choice of various screw caps.

 : Supplied without screw caps - please order separately.

 : Packaged in impermeable plastic packs.

Specifications
  • Capacity:
    150 ml
  • Seq:
    06